EMAIL THIS PAGE TO A FRIEND
Research in veterinary science
Cryopreservation of Trypanosoma evansi after DEAE-cellulose purification: Evaluation of infective parameters.
PMID 20542526
Abstract
Cryopreservation is a method of keeping parasites alive in a laboratory. However, this technique may also damage the parasite. Alternatively, parasites may be maintained by in vitro culture. Unfortunately, for Trypanosoma evansi no effective medium that is able to maintain the parasite for more than 4 months has been described. In this study, we examined the effect of purifying trypomastigote through DEAE-cellulose chromatography before and after cryopreservation, by analyzing the pre-patent period, longevity, parasitemia, and count of viable parasites. Our results showed a three-times increase in the concentration of viable trypomastigote in DEAE-purified cryopreserved parasites as compared to non-DEAE-purified cryopreserved parasites. This indicates that DEAE-cellulose chromatography followed by cryopreservation is an effective method for the storage and preservation of T. evansi, with the advantage that the stocked parasites will be ready to use in molecular biology procedures.