EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Novel coumarin derivatives as potential antidyslipidemic agents.
PMID 20542691
Abstract
A series of novel benzocoumarin derivatives were synthesized and evaluated for their in vivo antidyslipidemic and in vitro antioxidant activities. Among 11 compounds tested, 2 compounds showed potent antidyslipidemic activity and 3 compounds showed potent antioxidant activity.