EMAIL THIS PAGE TO A FRIEND
Brain research
The amnesic substance 2-deoxy-D-galactose suppresses the maintenance of hippocampal LTP.
PMID 2054616
Abstract
Male Wistar rats were intraventricularly injected with 2-deoxy-D-galactose (do-gal), a substance interfering with the fucosylation of glycomacromolecules and impairing memory consolidation in various learning tasks. Do-gal was found to have no influence on the monosynaptically evoked field potential (MEFP) recorded in the dentate gyrus upon stimulation of the perforant pathway. However, hippocampal long-term potentiation (LTP) induced in do-gal-pretreated animals by fractionated tetanization of the perforant pathway declined to control levels 2 h after tetanization, whereas it remained constant for 24 h in saline-treated rats. Similar effects were observed in the CA1 region of hippocampal slices. The results indicate a participation of fucosylated macromolecules in the maintenance of LTP. The possible significance of processes involved in LTP for memory formation is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4407
2-Deoxy-D-galactose, 98%
C6H12O5