EMAIL THIS PAGE TO A FRIEND
Maturitas
Effects of continuous combined hormone replacement therapy and clodronate on bone mineral density in osteoporotic postmenopausal women: a 5-year follow-up.
PMID 20547017
Abstract
To determine the effects of HRT with or without clodronate on bone mineral density (BMD) change and bone turnover markers. Prospective, partly randomized trial. Kuopio University Hospital, Finland. 167 osteoporotic women (61+/-2.7 years; T-score
Related Materials