EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Cloning, sequencing and expression analysis of a gene encoding alcohol oxidase in Paenibacillus sp. AIU 311.
PMID 20547358
Abstract
We have cloned a gene encoding an alcohol oxidase (AOD) specific to aldehyde alcohols from Paenibacillus sp. AIU 311. The AOD gene contains an open reading frame consisting of 618 nucleotides corresponding to 205 amino acid residues. The deduced amino acid sequence exhibits a high similarity to that of manganese superoxide dismutases (SODs). We expressed the cloned gene as an active product in Escherichia coli BL21 cells. The productivity (total units per culture broth volume) of the recombinant AOD expressed in E. coli BL21 is 26,000-fold higher than that of AOD in Paenibacillus sp. AIU 311. The recombinant AOD also exhibits aldehyde alcohol oxidase activity and SOD activity. The recombinant cells described in this study have utility for the production of glyoxal from glycolaldehyde.