EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Potential dermal exposure in greenhouses for manual sprayers: analysis of the mix/load, application and re-entry stages.
PMID 20547410
Abstract
An evaluation of the Potential Dermal Exposure for the mix/load, application and re-entry stages, associated with procymidone and deltamethrin usage, was carried out for tomatoes grown in greenhouses of small production units in Argentina. Eight experiments were done with four different operators, under typical field conditions with a lever operated backpack sprayer. The methodology applied was based on the Whole Body Dosimetry technique, evaluating a set of different data for the mix and load, application and re-entry operations. These results indicated that the Potential Dermal Exposure of the application step was (38+/-17)mLh(-1) with the highest proportion on torso, head and arms. When the three stages were compared, re-entry was found to contribute least towards the total Potential Dermal Exposure; meanwhile in all cases, except one, the mix/load operation was the stage with highest exposure. The Margin of Safety for each different operation was also calculated and the proportion of pesticide drift from the greenhouse to the environment is presented. These results emphasize the importance of improving the personal protection measures in the mix and load stage, an operation that is not usually associated with high-risk in small production units.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36640
Procymidone, PESTANAL®, analytical standard
C13H11Cl2NO2