EMAIL THIS PAGE TO A FRIEND
Archives of insect biochemistry and physiology
Purification and properties of trypsin-like enzyme from the midgut of Morimus funereus (coleoptera, cerambycidae) Larvae.
PMID 20549815
Abstract
Trypsin-like enzyme (TLE) from the anterior midgut of Morimus funereus larvae was purified by anion exchange chromatography and gel filtration chromatography and characterized. Specific TLE activity was increased 322-fold by purification of the crude midgut extract. The purified enzyme had a pH optimum of 9.0 (optimum pH range 8.5-9.5) and temperature optimum of 45 degrees C with the K(M) ratio of 0.065 mM for benzoyl-arginine-p-nitroanilide (BApNA). Among a number of inhibitors tested, the most efficient was benzamidine (K(I) value of 0.012 mM, Ic(50) value of 0.204 mM) while inhibition of TLE activity by SBTI, TLCK, and PMSF was partial. Almost all divalent cations tested enhanced the enzyme activity, amongst them Co2+ and Mn2+ stimulated TLE activity for 2.5 times. The purified TLE (after gel-filtration on Superose 12 column) had a molecular mass of 37.5 kDa with an isoelectric point over 9.3. Sodium dodecylsulphate-polyacrylamide gel electrophoresis (SDS-PAGE) revealed one band of 38 kDa, suggesting that the enzyme is a monomer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
434760
Benzamidine hydrochloride, 99%
C7H8N2 · HCl
63226
Benzamidine hydrochloride 1 M solution