EMAIL THIS PAGE TO A FRIEND
Natural product research
Alkaloids from Glaucium flavum from Sardinia.
PMID 20552526
Abstract
Glaucium flavum collected in Sardinia was studied using a phytochemical approach in order to evaluate its alkaloid composition and obtain a comparison with the alkaloid contents of the same species in populations of other geographic proveniences. In fact, different chemoecotypes of G. flavum have been identified, on the basis of their particular content and composition in alkaloids, in accordance with the different distribution areas. The analysis showed that Sardinian G. flavum contains a homogeneous alkaloid pattern of aporphyne type, significantly different from those reported for populations from other parts of Europe.