EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Stimulus-dependent requirement of ion channels for microglial NADPH oxidase-mediated production of reactive oxygen species.
PMID 20554029
Abstract
Reactive oxygen species (ROS) produced by activated microglial cells play a pivotal role in the pathogenesis of neuro-degenerative and neuro-inflammatory diseases. Here we demonstrate that the pro-inflammatory lipid lysophosphatidylcholine (LPC) is capable of inducing microglial ROS production, which is mediated by the activity of NADPH oxidase. Inhibition of TRPV1 non-selective cation channels abolished ROS production in LPC-stimulated microglia, whereas inhibitors of K(+) channels, H(+) channels and Cl(-) channels had no significant effects. In contrast, activity of all four ion channel types was required for PMA-induced NADPH oxidase-mediated ROS generation, suggesting a differential, stimulus-dependent regulation of microglial ROS production by ion channel activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9143
Phorbol 12,13-diacetate, powder
C24H32O8