EMAIL THIS PAGE TO A FRIEND
Toxicology and industrial health
Apoptosis contributes to testicular toxicity induced by two isomers of bromopropanes.
PMID 20554630
Abstract
The aim of this study was to investigate the different testicular toxicity and the role of apoptosis in the possible mechanism induced by the two isomers of bromopropanes (BPs) in the same dosage. Following the 14-day treatment with a single dose of 1-BP and 2-BP (1 g/kg), male rats were killed and a series of experiments were performed. 1-BP and 2-BP both significantly decreased the epididymal sperm count, while only 2-BP induced an increase in sperms with abnormal heads. Morphological evaluation showed that 1-BP did not cause morphological changes in seminiferous epithelium, but 2-BP treatment resulted in the disappearance of spermatogonia, atrophy of the seminiferous tubules and degeneration of germ cells. 2-BP significantly increased the TUNEL-positive cells and the activation of caspase-3 and decreased the genes and proteins expression of Bax, Bcl-2 and p53. In contrast, there were no significant changes in the expression of apoptosis-related genes and proteins in 1-BP group, though the TUNEL-positive cells were significantly increased. Taken together, this study indicated that those two isomers both have toxicity in male rats, however, the testicular toxicity and the role of apoptosis in the toxic mechanism induced by 1-BP and 2-BP may be different.
Related Materials