EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Construction of strong alkaline microcavities for facile synthesis of fluorescence-tunable ZnO quantum dots.
PMID 20556299
Abstract
Strong alkaline microcavities were constructed through the water-induced protonation of dodecylamine for the synthesis of fluorescence-tunable ZnO QDs in neutral bulk reaction solution.
Related Materials