EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Glucose metabolism activation by SHIP2 inhibitors via up-regulation of GLUT1 gene in L6 myotubes.
PMID 20558154
Abstract
Lipid phosphatase SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) plays an important role in the regulation of insulin signaling. In this report, we identified AS1938909, a novel small-molecule SHIP2 inhibitor. AS1938909 showed potent inhibition of SHIP2 (Ki=0.44 microuM) and significant selectivity over other related phosphatases. Further, AS1938909 increased Akt phosphorylation, glucose consumption, and glucose uptake in L6 myotubes. Treatment of L6 myotubes with SHIP2 inhibitors for 48 h significantly induced expression of GLUT1 mRNA, but not that of GLUT4. These results suggest that pharmacological inhibition of SHIP2 activates glucose metabolism due, at least in part, to up-regulation of GLUT1 gene expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1022 AS1949490, ≥98% (HPLC)
C20H18ClNO2S