EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Hydrophilicity, the major determining factor influencing the solvation environment of protic ionic liquids.
PMID 20563334
Abstract
The study of solubility of 4-nitroaniline and its N,N-diethyl derivative in ionic liquids (ILs) of varying water content has shown that it is the interaction between water and the ILs' ions, and not with the solute, that is the most important factor determining the solute's preferential hydration parameter and thus the solvation shell structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31569
4-Nitroaniline, analytical standard
C6H6N2O2