EMAIL THIS PAGE TO A FRIEND
Respiratory research
The role of pleiotrophin and beta-catenin in fetal lung development.
PMID 20565841
Abstract
Mammalian lung development is a complex biological process, which is temporally and spatially regulated by growth factors, hormones, and extracellular matrix proteins. Abnormal changes of these molecules often lead to impaired lung development, and thus pulmonary diseases. Epithelial-mesenchymal interactions are crucial for fetal lung development. This paper reviews two interconnected pathways, pleiotrophin and Wnt/beta-catenin, which are involved in fibroblast and epithelial cell communication during fetal lung development.
Related Materials