EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Nanoparticulate copper--routes towards oxidative stability.
PMID 20567754
Abstract
A modified polyol-based reduction method in ethylene glycol that incorporates poly(N-vinylpyrrolidone) (PVP, M(av) = 10,000; 40,000; 55,000) as polymeric anti-agglomerant alongside a reducing additive (N(2)H(4) x H(2)O, NaBH(4), NaH(2)PO(2) x H(2)O) has been employed to investigate the influence of synthetic parameters on the purity, morphology and stability of an array of polymer-coated copper nanoparticles. While data point to ethylene glycol being capable of acting as a reductant in this system, the use of NaH(2)PO(2) x H(2)O as co-reductant in tandem with the presence of PVP (M(av) 40,000) has rendered nanoparticles with a mean size distribution of 9.6 +/- 1.0 nm that exhibit stability towards oxidation for several months. These data allow us to probe fundamentally how oxidatively stable nano-copper might be achieved.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V3409
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%
C6H9NO