EMAIL THIS PAGE TO A FRIEND
Organic letters
A new synthesis of 2,3-di- or 2,3,3-trisubstituted 2,3-dihydro-4-pyridones by reaction of 3-ethoxycyclobutanones and N-p-toluenesulfonyl imines using titanium(IV) chloride: synthesis of (+/-)-bremazocine.
PMID 20568828
Abstract
N-p-Toluenesulfonyl (Ts) aldimines reacted with 3-ethoxycyclobutanones by catalysis of titanium(IV) chloride to afford 2,3-di- or 2,3,3-trisubstituted N-Ts-2,3-dihydro-4-pyridones. Synthesis of (+/-)-bremazocine was efficiently accomplished by using this method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14010
Titanium(III) chloride solution, ≥12% TiCl3 basis
Cl3Ti