EMAIL THIS PAGE TO A FRIEND
Critical reviews in toxicology
Genetic toxicology of folpet and captan.
PMID 20569196
Abstract
Folpet and captan are fungicides whose genotoxicity depends on their chemical reaction with thiols. Multiple mutagenicity tests have been conducted on these compounds due to their positive activity in vitro and their association with gastrointestinal tumors in mice. A review of the collective data shows that these compounds have in vitro mutagenic activity but are not genotoxic in vivo. This dichotomy is primarily due to the rapid degradation of folpet and captan in the presence of thiol-rich matrices typically found in vivo. Genotoxicity has not been found in the duodenum, the mouse tumor target tissue. It is concluded that folpet like captan presents an unlikely risk of genotoxic effects in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32054
Captan, PESTANAL®, analytical standard
C9H8Cl3NO2S
32057
Folpet, PESTANAL®, analytical standard
C9H4Cl3NO2S