EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Minor crops for export: a case study of boscalid, pyraclostrobin, lufenuron and lambda-cyhalothrin residue levels on green beans and spring onions in Egypt.
PMID 20574869
Abstract
Dissipation rates of boscalid [2-chloro-N-(4' -chlorobiphenyl-2-yl)nicotinamide], pyraclostrobin [methyl 2-[1-(4-chlorophenyl) pyrazol-3-yloxymethyl]-N-methoxycarbanilate], lufenuron [(RS)-1-[2,5-dichloro-4-(1,1,2,3,3,3-hexafluoropropoxy)phenyl]-3-(2,6-difluorobenzoyl)urea] and lambda-cyhalothrin [(R)-cyano(3-phenoxyphenyl)methyl (1S,3S)-rel-3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylate] in green beans and spring onions under Egyptian field conditions were studied. Field trials were carried out in 2008 in a Blue Nile farm, located at 70 kilometer (km) from Cairo (Egypt). The pesticides were sprayed at the recommended rate and samples were collected at pre-determined intervals. After treatment (T(0)) the pesticide residues in green beans were 7 times lower than in spring onions. This is due to a different structure of vegetable plant in the two crops. In spring onions, half-life (t(1/2)) of pyraclostrobin and lufenuron was 3.1 days and 9.8 days respectively. At day 14th (T(14)) after treatment boscalid residues were below the Maximum Residue Limit (MRL) (0.34 versus 0.5 mg/kg), pyraclostrobin and lambda -cyhalothrin residues were not detectable (ND), while lufenuron residues were above the MRL (0.06 versus 0.02 mg/kg). In green beans, at T(0), levels of boscalid, lufenuron and lambda -cyhalothrin were below the MRL (0.28 versus 2 mg/kg; ND versus 0.02 mg/kg; 0.06 versus 0.2 mg/kg, respectively) while, after 7 days treatment (T(7)) pyraclostrobin residues were above the MRL (0.03 versus 0.02 mg/kg). However, after 14 days the residue level could go below the MRL (0.02 mg/kg), as observed in spring onions.