EMAIL THIS PAGE TO A FRIEND
Journal of biomedical materials research. Part B, Applied biomaterials
Tungsten-rhenium suture needles with improved properties for coronary artery bypass graft surgery.
PMID 20574974
Abstract
Suture needles are essential instruments for performing blood vessel anastomosis in coronary artery bypass graft (CABG) surgeries. Exceptional needle performance is needed now as never before because of the increasing prevalence of elderly patients with advanced stages of coronary disease and calcified tissue. The various properties that affect the performance of suture needles used in CABG surgery are reviewed and recent progress towards improving needle performance through the use of tungsten-rhenium alloys and novel lubricous needle coatings is presented. Substantial enhancement of properties beyond those exhibited by commercially available stainless steel suture needles has been made, including an approximate 40% increase in strength, 100% increase in stiffness, and superior multiple pass penetration performance in both synthetic rubber media and human cadaver carotid arteries (p < 0.01).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459984
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis
Re
267317
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Re
204188
Rhenium, powder, 99.995% trace metals basis
Re
267279
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Re
GF57863931
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Re
GF60560513
Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%
Re
GF90374441
Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%
Re
GF77806367
Rhenium, wire reel, 0.1m, diameter 0.5mm, annealed, 99.97%
Re
GF63991929
Rhenium, wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%
Re
GF14519156
Rhenium, wire reel, 0.2m, diameter 0.076mm, annealed, 99.97%
Re
GF59830139
Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Re
GF96591875
Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%
Re
GF02183217
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Re
GF70064658
Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%
Re
GF32991486
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Re
GF48358435
Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%
Re
GF96805884
Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%
Re
GF25796010
Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%
Re
GF98990050
Rhenium, wire reel, 0.2m, diameter 1.0mm, annealed, 99.97%
Re
GF14894163
Rhenium, wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%
Re
GF78766745
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Re
GF73984797
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Re
GF72901324
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Re
GF58081945
Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%
Re
GF23936104
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Re
GF44804325
Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%
Re
GF00735353
Rhenium, wire reel, 0.5m, diameter 0.3mm, annealed, 99.97%
Re
GF24502115
Rhenium, wire reel, 0.5m, diameter 0.5mm, annealed, 99.97%
Re
GF61770225
Rhenium, wire reel, 1m, diameter 0.076mm, annealed, 99.97%
Re
GF45842733
Rhenium, wire reel, 1m, diameter 0.10mm, annealed, 99.97%
Re