EMAIL THIS PAGE TO A FRIEND
European journal of heart failure
Loop diuretic therapy in left ventricular systolic dysfunction: has familiarity bred contempt for a critical but potentially nephrotoxic cardio renal therapy?
PMID 20576834