EMAIL THIS PAGE TO A FRIEND
Drug development and industrial pharmacy
The influence of lipid characteristics on the formation, in vitro release, and in vivo absorption of protein-loaded SLN prepared by the double emulsion process.
PMID 20578879
Abstract
To study the influence of lipid characteristics on the formation, in vitro release, and in vivo absorption of solid lipid nanoparticles (SLN) prepared by the double emulsion method. Stearic acid (SA), octadecyl alcohol (OA), cetyl palmitate (CP), glyceryl monostearate (GM), glyceryl palmitostearate (GP), glyceryl tripalmitate (GT), and glyceryl behenate (GB) were selected as the representatives of different kinds of lipids, insulin and thymopentin (TP5) were selected as the model protein drugs. Before preparation, the contact angles between water and lipids were determined to investigate their hydrophobicity. The influence of lipid hydrophobicity or lipid solution viscosity on the preparation of primary emulsion, double emulsion, and SLN were studied by evaluating the particle size, state, and stability of the systems. CP-SLN, GT-SLN, and GP-SLN were selected to be loaded with insulin and TP5 for the in vitro release and in vivo absorption examination. After oral administration to diabetic rats, the pharmacological availability (PA) of insulin-CP-SLN, insulin-GP-SLN, and insulin-GT-SLN were determined. The hydrophobicity order of the lipids was GM
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8806
Thymopentin, ≥97% (HPLC)
C30H49N9O9