EMAIL THIS PAGE TO A FRIEND
Dermatology online journal
Generalized skin drug eruption to natalizumab in a patient with multiple sclerosis.
PMID 20579469
Abstract
We report a generalized skin eruption in a young man being treated with natalizumab, a new drug used in patients with multiple sclerosis.