EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Determination of non-steroidal anti-inflammatory drugs residues in animal muscles by liquid chromatography-tandem mass spectrometry.
PMID 20579495
Abstract
A confirmatory method for the determination of residues of nine non-steroidal anti-inflammatory drugs and one metabolite in animal muscles has been developed. After enzymatic hydrolysis samples were extracted with acetonitrile and cleaned up using alumina and C(18) SPE cartridges. Liquid chromatography-tandem mass spectrometry was used for the separation and determination of analytes. The method was validated in bovine muscles, according to the Commission Decision 2002/657/EC criteria. Applicability of the method in the analysis of swine, horse and chicken muscles was checked by precision and recovery experiment. The influence of matrix effect on the quantification of non-steroidal anti-inflammatory drugs residues was investigated. The method was used for the confirmation of phenylbutazone and oxyphenbutazone in horse muscle sample.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0540 Oxyphenbutazone, ≥98% (HPLC)
C19H20N2O3