EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Modular synthesis of 1,2-diamine derivatives by palladium-catalyzed aerobic oxidative cyclization of allylic sulfamides.
PMID 20583013
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
299685
Palladium(II) trifluoroacetate, 97%
C4F6O4Pd