EMAIL THIS PAGE TO A FRIEND
Agents and actions
Toxicological properties of carrageenan.
PMID 2058470
Abstract
Food-grade carrageenan is a safe natural product prepared from seaweed. Its addition to food imparts many desirable characteristics which have allowed it to be used continuously for centuries. The long safe history of this natural food additive is confirmed by negative results in subchronic and chronic feeding studies in many animal species, mutagenicity studies and reproductive toxicity studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1013 Carrageenan, suitable for gel preparation