EMAIL THIS PAGE TO A FRIEND
The Analyst
Antibody functionalized interdigitated micro-electrode (IDmicroE) based impedimetric cortisol biosensor.
PMID 20589269
Abstract
This paper reports on an ultrasensitive, disposable, impedimetric biosensor for cortisol detection. C-Mab (a Cortisol specific monoclonal antibody) was covalently immobilized via amide bond on the surface of the interdigitated micro-electrodes (IDmicroEs) functionalized with dithiobis(succinimidyl propionate) (DTSP) self-assembled monolayer (SAM). After C-Mab binding, unreacted active groups of DTSP were blocked using ethanolamine (EA) and glycine (Gly) mixture. The disposable sensors were exposed to solutions with different cortisol concentrations and a label-free electrochemical impedance (EIS) technique was used to determine the cortisol concentration. EIS results confirm that the EA-Gly/C-Mab/DTSP/IDmicroE based biosensor exhibited the sensitivity of 2.855 kohms M(-1) and could accurately detect cortisol in the range of 1 pM to 10 nM in saliva. This work establishes the feasibility of using an impedance based biosensor as a disposable cortisol detector, capable of working with complex bodily fluids (e.g., saliva). The architecture enables the use of cortisol sensors at point-of-care.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3669
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder
C14H16N2O8S2