EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Highly efficient and facile synthesis of 5-azido-2'-deoxyuridine.
PMID 20589573
Abstract
Previously reported syntheses of the photoaffinity label 5-azido-2'-deoxyuridine are rather inefficient and involve the tedious preparation of a 5-nitro intermediate. To overcome these inconveniences, we have developed a new approach from the commercially available 5-bromo-2'-deoxyuridine nucleoside. Our synthetic route makes use of a benzylamination reduction sequence. Using this strategy, the 5-azido-2'-deoxyuridine photolabel is prepared in three steps and quantitative yields.