EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Discovery process and characterization of novel carbohydrazide derivatives as potent and selective GHSR1a antagonists.
PMID 20593439
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C11006
Carbohydrazide, 98%
CH6N4O