EMAIL THIS PAGE TO A FRIEND
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology
[Safety measure against improper use of lime sulfur].
PMID 20593653
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
409553
Calcium sulfide, −325 mesh, ≥99.9% trace metals basis
CaS