EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis in a girl due to several cosmetics containing diazolidinyl-urea or imidazolidinyl-urea.
PMID 20597934
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5146
Diazolidinyl urea, ≥95%
C8H14N4O7
I5133
Imidazolidinyl urea
C11H16N8O8