EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience methods
Assaying multiple biochemical variables from the same tissue sample.
PMID 20600314
Abstract
Experiments often involve multiple analyses, such as assays of neurotransmitters and proteins, and this can require different initial sample preparations. Typically, this is accomplished by using different animals or different tissue samples from the same animal. Either approach renders comparisons between assays more variable and greatly increases the effort and/or cost. Using tissue collected from rat striatum and molecules of special relevance to studies of Parkinson's disease, we show that tissue sonication in water prior to aliquoting into the appropriate concentrated solutions (e.g. HClO(4) and lysis buffers) permits several types of measurements to be made from the same initial samples. Dopamine and its metabolite homovanillic acid, serotonin and its metabolite 5-hydroxyindoleacetic acid, tyrosine hydroxylase and its phosphorylation at Ser19 and Ser31, and the dopamine transporter were unaffected. However, phospho-Akt levels fell slightly and phospho-ERK1/2 tended to drop. We also present a simple technique to preserve phosphorylation state of proteins such as ERK1/2 by perfusing animals through the heart with a phosphatase inhibitor, NaF. Dopamine metabolite dihydroxyphenyl acetic acid (DOPAC) levels were raised with both techniques, however. The general principles reported here are likely to apply to other brain regions, facilitate multiple comparisons of variables, increase efficiency, and decrease costs.