EMAIL THIS PAGE TO A FRIEND
Danish medical bulletin
Pharmacokinetics and pharmacodynamics of disopyramide.
PMID 2060320
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2920000
Disopyramide, European Pharmacopoeia (EP) Reference Standard
C21H29N3O