EMAIL THIS PAGE TO A FRIEND
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan
Analytical method for tributyltin and triphenyltin contained in household products-preparing for the revision of authorized analytical method-.
PMID 20606374
Abstract
In preparing for the revision of the authorized analytical method for tributyltin (TBT) and triphenyltin (TPT), which are banned from using according to the "Act on the Control of Household Products Containing Harmful Substances", an examination was conducted on the detection method of these substances using gas chromatography/mass spectrometry (GC/MS), after derivatizing them (ethyl-derivatizing method and hydrogen-derivatizing method). Ethyl-derivatized compounds had stability, which enabled the detection of TPT with a higher sensitivity. In addition, a preparation suitable for the following analytical objects was established: (1) textile products, (2) water-based products (such as water-based paint), (3) oil-based products (such as wax), and (4) adhesives. Addition-recovery experiments were conducted using the prescribed pretreatment method, when each surrogate substances (TBT-d27, TPT-d15) were added and the data were corrected, good recovery rates (94.5-118.6% in TBT, and 86.6-110.1% in TPT) were obtained. When TBT and TPT in 31 commercially available products were analyzed based on the developed analytical method, an adhesive showed 13.2 microg/g of TBT content, which exceeded the regulatory criterion (1 microg/g as tin). Next, when the same products with different manufacturing date were analyzed, TBT (10.2-10.8 microg/g), which exceeded the regulatory criterion, was detected in 4 products among 8 products, and simultaneously, a high concentration (over 1000 microg/g) of dibutyltin (DBT) was detected. It was suggested that TBT as an impurity of DBT remained, and the manufacturer chose the voluntary recall of the product. The new method is considered sufficiently applicable as a revised method for the conventionally authorized method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
244775
Triphenyltin hydride
C18H16Sn