EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
A general palladium-catalyzed sonogashira coupling of aryl and heteroaryl tosylates.
PMID 20607775
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
299685
Palladium(II) trifluoroacetate, 97%
C4F6O4Pd