EMAIL THIS PAGE TO A FRIEND
Pigment cell & melanoma research
Gamma- and delta-tocotrienols inhibit skin melanin synthesis by suppressing constitutive and UV-induced tyrosinase activation.
PMID 20609176
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0577 D-δ-Tocotrienol, from rice, ≥65% (GC)
C27H40O2
69745
δ-Tocotrienol, analytical standard
C27H40O2