EMAIL THIS PAGE TO A FRIEND
Journal of the Royal Society of Medicine
Music and matrimony--hazards for the colophonium allergic patient.
PMID 20610620
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60895 Gum rosin, natural resin