EMAIL THIS PAGE TO A FRIEND
Anesthesiology
Spinal alpha 2-adrenoceptor-mediated analgesia in neuropathic pain reflects brain-derived nerve growth factor and changes in spinal cholinergic neuronal function.
PMID 20613480