EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Immunoadsorbents for clinical use: ex vivo immunoglobulin E removal in allergy.
PMID 2061384
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CL4B200 Sepharose® CL-4B, Cross-linked