EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Peptide deformylase inhibitors with retro-amide scaffold: synthesis and structure-activity relationships.
PMID 20615695
Abstract
Peptide deformylase (PDF) is a metalloprotease catalyzing the removal of a formyl group from newly synthesized proteins. Thus inhibition of PDF activity is considered to be one of the most effective antibiotic strategies. Reported herein are the synthesis and structure-activity relationship studies of retro-amide inhibitors based on actinonin, a naturally occurring PDF inhibitor. Analysis of the structure-activity relationships led to the discovery of 7a, which exhibits potent enzyme inhibition and antibacterial activity against Streptococcus pneumoniae, Haemophilus influenzae, and Moraxella catarrhalis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6671
Actinonin
C19H35N3O5