EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Degradation of acid fuchsine by a modified electro-Fenton system with magnetic stirring as oxygen supplying.
PMID 20617731
Abstract
The current modified electro-Fenton system was designed to develop a more convenient and efficient undivided system for practical wastewater treatment. The system adopted a cathode portion that employed magnetic stirring instead of common oxygen gas diffusion or gas sparging to supply oxygen gas for the electrolyte solution. Key factors influencing the cathode fabrication and activity were investigated. The degradation of acid fuchsine with a self-made graphite-polytetrafluorethylene cathode was studied using a spectrophotometer. It was found that the cathode generated hydrogen peroxide with high current efficiency and the hydrogen peroxide yield of the cathode did not decay after 10 times reuse. With the Pt anode at a ferrous ion concentration of 0.5 mmol/L, a pH of 3, and using magnetic stirring, dye decolorization could be rapidly accomplished but the destruction of benzene rings and intermediates was fairly difficult. With a Fe anode, dye degradation was more complete.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8129
Acid Fuchsin, used in tissue staining
C20H17N3Na2O9S3