EMAIL THIS PAGE TO A FRIEND
Cancer letters
S-trityl-L-cysteine derivative induces caspase-independent cell death in K562 human chronic myeloid leukemia cell line.
PMID 20619960
Abstract
Effect of CF(3)-STLC, a potent kinesin spindle protein (KSP) inhibitor, on K562 human CML cell line was investigated. Treatment with CF(3)-STLC induced mitotic arrest of the cell cycle with the appearance of characteristic monoastral spindles, subsequent apoptotic cell death and cleavage of PARP-1, caspase-3, and 4E-BP1. The wide ranging caspase inhibitor z-VAD fmk prevented the cleavage of caspase-3 and 4E-BP1, but failed to attenuate PARP-1 cleavage or cell death triggered by CF(3)-STLC. These results suggest that CF(3)-STLC can induce apoptotic cell death in a caspase-independent manner, and may work effectively as an anti-cancer agent for hematological malignancies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
164739
(+)-S-Trityl-L-cysteine, 97%
C22H21NO2S