EMAIL THIS PAGE TO A FRIEND
Gastrointestinal endoscopy
An interesting case of iron deficiency anemia.
PMID 20620279
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45667
2,4,5-T, PESTANAL®, analytical standard
C8H5Cl3O3