EMAIL THIS PAGE TO A FRIEND
Preparative biochemistry & biotechnology
1,6-Hexanediamine methanesulfonate: a mild and efficient catalyst for the tetrahydropyranylation of alcohols under solvent-free conditions.
PMID 20623428
Abstract
Various alcohols react with 3,4-dihydro-2 H-pyran under mild conditions using a catalytic amount of 1,6-hexanediamine methanesulfonate. It affords the corresponding tetrahydropyranyl ethers in good yields at a faster rate in the absence of solvent.
Related Materials