EMAIL THIS PAGE TO A FRIEND
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology
[Effectiveness of combined application of ursosan and anxiolytic 2-mercaptobenzimidazole in early stage of cholelithiasis].
PMID 20623958
Abstract
We spent the complex investigation of therapeutic efectiveness of ursosan and afobasol in 125 patients with gallstone disease on early stage (before stones). After the course of treatment we revealed positive changes of clinical symptoms and improvement of biochemical bile compound. Besides we found the positive changes in hormone levels correlated with decreasing of bile lithogenity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3205
2-Mercaptobenzimidazole, 98%
C7H6N2S