EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Phenoxazine based units--synthesis, photophysics and electrochemistry.
PMID 20625802
Abstract
A few new phenoxazine-based conjugated monomers were synthesized, characterized, and successfully used as semiconducting materials. The phenoxazine-based oligomers have low ionization potentials or high-lying HOMO levels (~4.7 eV), which were estimated from cyclic voltammetry. Conjugated oligomers offer good film-forming, mechanical and optical properties connected with their wide application. These results demonstrate that phenoxazine-based conjugated mers are a promising type of semiconducting and luminescent structures able to be used as thin films in organic electronics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
414506
2-(Tributylstannyl)furan, 97%
C16H30OSn
414492
2-(Tributylstannyl)thiophene, 97%
C16H30SSn