EMAIL THIS PAGE TO A FRIEND
International journal of obstetric anesthesia
Side effects of spiramycin masquerading as local anesthetic toxicity during labor epidural analgesia.
PMID 20627689
Abstract
Significant fetal bradycardia occurred when a parturient receiving labor epidural analgesia experienced generalized numbness and tingling, a metallic taste and hot flushes. An emergent cesarean delivery under general anesthesia was performed with favorable outcomes for the mother and baby. The most likely source of the maternal symptoms was spiramycin, which was being administered for treatment of toxoplasmosis.