EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Voltage-gated divalent currents in descending vasa recta pericytes.
PMID 20630935
Abstract
Multiple voltage-gated Ca(2+) channel (Ca(V)) subtypes have been reported to participate in control of the juxtamedullary glomerular arterioles of the kidney. Using the patch-clamp technique, we examined whole cell Ca(V) currents of pericytes that contract descending vasa recta (DVR). The dihydropyridine Ca(V) agonist FPL64176 (FPL) stimulated inward Ca(2+) and Ba(2+) currents that activated with threshold depolarizations to -40 mV and maximized between -20 and -10 mV. These currents were blocked by nifedipine (1 μM) and Ni(2+) (100 and 1,000 μM), exhibited slow inactivation, and conducted Ba(2+) > Ca(2+) at a ratio of 2.3:1, consistent with "long-lasting" L-type Ca(V). In FPL, with 1 mM Ca(2+) as charge carrier, Boltzmann fits yielded half-maximal activation potential (V(1/2)) and slope factors of -57.9 mV and 11.0 for inactivation and -33.3 mV and 4.4 for activation. In the absence of FPL stimulation, higher concentrations of divalent charge carriers were needed to measure basal currents. In 10 mM Ba(2+), pericyte Ca(V) currents activated with threshold depolarizations to -30 mV, were blocked by nifedipine, exhibited voltage-dependent block by diltiazem (10 μM), and conducted Ba(2+) > Ca(2+) at a ratio of ∼2:1. In Ca(2+), Boltzmann fits to the data yielded V(1/2) and slope factors of -39.6 mV and 10.0 for inactivation and 2.8 mV and 7.7 for activation. In Ba(2+), V(1/2) and slope factors were -29.2 mV and 9.2 for inactivation and -5.6 mV and 6.1 for activation. Neither calciseptine (10 nM), mibefradil (1 μM), nor ω-agatoxin IVA (20 and 100 nM) blocked basal Ba(2+) currents. Calciseptine (10 nM) and mibefradil (1 μM) also failed to reverse ANG II-induced DVR vasoconstriction, although raising mibefradil concentration to 10 μM was partially effective. We conclude that DVR pericytes predominantly express voltage-gated divalent currents that are carried by L-type channels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F131
FPL 64176, ≥98% (HPLC), powder
C22H21NO3