EMAIL THIS PAGE TO A FRIEND
Journal of acupuncture and meridian studies
Treatment of Mycoplasma wenyonii infection in cows with imidocarb dipropionate injection-acupuncture.
PMID 20633467
Abstract
Sixteen shorthorn cows from Xiazhuang farm were admitted to the Veterinary Teaching Hospital at the College of Animal Science and Veterinary Medicine, Shandong Agricultural University for evaluation of poor appetite, listlessness, fever, tachycardia, tachypnea, lethargy, positive jugular venous pulse and anemia. Blood smear examination and polymerase chain reaction analysis in these cows revealed an infection with Mycoplasma wenyonii. The subjects were divided into two groups: control group (three cows) treated with intramuscular injection with imidocarb dipropionate (3 mg/kg/day for 2 days) and the experimental group (13 cows), treated with injection-acupuncture (Imidocarb Dipropionate, 1 mg/kg, once every 3 days for 6 days) at BL17, BL18, BL20, BL25, ST36, SP06 and CV04. At day 15, negative results were found using blood smear examination in all control and experimental groups.
Related Materials