EMAIL THIS PAGE TO A FRIEND
Journal of natural medicines
New iridoid diesters of glucopyranose from Linaria canadensis (L.) Dum.
PMID 20635154
Abstract
Two new iridoid diesters of glucopyranose were isolated from the aerial part of Linaria canadensis (L.) Dum. Eight known flavones, apigenin, diosmetin, genkwanin, luteolin, luteolin 7-O-glucoside, luteolin 7-O-glucuronide, genkwanin 4'-O-rutinoside, and quercetin 7-O-rutinoside were also isolated. The chemical structures of the isolated compounds were elucidated based on the analyses of the spectroscopic data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7321
Diosmetin
C16H12O6
90985
Diosmetin, analytical standard
C16H12O6