EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Identification of chalcones as in vivo liver monofunctional phase II enzymes inducers.
PMID 20639112
Abstract
Cancer preventive agents (CPA) are drugs able to suppress the carcinogen metabolic activation or block the formation of ultimate carcinogens. CPA could act through various molecular mechanisms, for example by interfering with the action of procarcinogen. This could be attained by increasing the phase II enzymes levels of quinone reductase (QR) and glutathione S-transferase (GST). New flavonoids, especially chalcones, have been identified as in vivo monofunctional phase II enzymes inducers. Oral administration of chalcone, 4, and both p-methoxy-substituted chalcones, 6 and 14, increased hepatic QR activity with concomitant decrease in CYP1A1 activity, a member of the most important group of phase I enzymes cytochrome P450. Among them, 4 also increased GST activity. While p-bromo-substituted chalcone 8 was the best inducer of QR it decreased hepatic GST expression and cytochrome P450, being the most effective decreasing cytochrome P450-expression. Thienyl-chalcone 20 being the bioisostere of chalcone 4 did not display the same in vivo profile in the phase I level modification. As chalcone 4 its bioisostere, chalcone 20, displayed low DNA strand breakage and absence of mutagenicity. Also, in our preliminary in vivo tumourigenesis/chemopreventive and acute-toxicity studies, chalcones 4, 6 and 8 showed the best behaviours as CPA justifying additional studies that are ongoing.